siRNA Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007010.1 | - | 1030 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX516018.1 | - | 1 | 381 | Chr. 15 - 38519438 - 38519940 [+] on Build GRCm38 | |

Azin1
Genomic Map