siRNA Details
| Gene Symbol: Gm41043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029426.1 | - | 2 | 504 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614247.1 | - | 2 | 504 | Chr. 13 - 101544396 - 101548152 [-] on Build GRCm38 | |

Gm41043
Genomic Map