siRNA Details
| Gene Symbol: Gm15527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080009.1 | - | 810 | |||
| AK146634.1 | - | 4 | 808 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15527| Gene Symbol: Gm15527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080009.1 | - | 810 | |||
| AK146634.1 | - | 4 | 808 | |||