siRNA Details
| Gene Symbol: 1700092E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045933.1 | 1 | 249 | |||
| GenBank mRNA | AK007041.1 | - | 1 | 244 | ||
| AK133453.1 | - | 1 | 253 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617294.1 | - | 1 | 253 | Chr. 13 - 26284151 - 26312410 [-] on Build GRCm38 | |

1700092E19Rik
Genomic Map