siRNA Details
| Gene Symbol: 1700092E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045933.1 | 3 | 1278 | |||
| GenBank mRNA | AK133453.1 | - | 3 | 1282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700092E19Rik| Gene Symbol: 1700092E19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045933.1 | 3 | 1278 | |||
| GenBank mRNA | AK133453.1 | - | 3 | 1282 | ||