siRNA Details
| Gene Symbol: 6330563C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018239.1 | - | 1 | 856 | ||
| AK047772.1 | - | 1 | 818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330563C09Rik| Gene Symbol: 6330563C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018239.1 | - | 1 | 856 | ||
| AK047772.1 | - | 1 | 818 | |||