siRNA Details
| Gene Symbol: Gm15133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040749.1 | 7 | 743 | |||
| GenBank mRNA | AK085986.1 | - | 7 | 744 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB519837.2 | - | 5 | 563 | Chr. 7 - 104317642 - 104324825 [+] on Build GRCm38 | |

Gm15133
Genomic Map