siRNA Details
| Gene Symbol: Gm15133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040749.1 | 1, 2 | 71 | |||
| GenBank mRNA | AK085986.1 | - | 1, 2 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15133| Gene Symbol: Gm15133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040749.1 | 1, 2 | 71 | |||
| GenBank mRNA | AK085986.1 | - | 1, 2 | 72 | ||