siRNA Details
| Gene Symbol: 9230118N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020357.1 | - | 1 | 494 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721170.1 | - | 1 | 490 | Chr. 2 - 152104678 - 152105205 [-] on Build GRCm38 | |

9230118N01Rik
Genomic Map