siRNA Details
| Gene Symbol: 9430019J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040635.1 | 3 | 901 | |||
| GenBank mRNA | AK020426.1 | - | 3 | 902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430019J16Rik| Gene Symbol: 9430019J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040635.1 | 3 | 901 | |||
| GenBank mRNA | AK020426.1 | - | 3 | 902 | ||