siRNA Details
| Gene Symbol: 9430019J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040635.1 | 3 | 320 | |||
| GenBank mRNA | AK020426.1 | - | 3 | 321 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721614.1 | - | 3 | 317 | Chr. 2 - 75436329 - 75444176 [-] on Build GRCm38 | |

9430019J16Rik
Genomic Map