siRNA Details
| Gene Symbol: 1700052I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033786.1 | 1 | 201 | |||
| GenBank mRNA | AK006769.1 | - | 1 | 171 | ||
| AK018897.1 | - | 1 | 172 | |||
| AK018974.1 | - | 1 | 203 | |||
| BY707327.1 | - | 1 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700052I22Rik| Gene Symbol: 1700052I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033786.1 | 1 | 201 | |||
| GenBank mRNA | AK006769.1 | - | 1 | 171 | ||
| AK018897.1 | - | 1 | 172 | |||
| AK018974.1 | - | 1 | 203 | |||
| BY707327.1 | - | 1 | 202 | |||