siRNA Details
| Gene Symbol: 1700051K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006761.1 | - | 1 | 60 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY764403.1 | - | 1 | 172 | Chr. 6 - 134245768 - 134246478 [-] on Build GRCm38 | |

1700051K13Rik
Genomic Map