siRNA Details
| Gene Symbol: 1700072O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045733.1 | 4 | 630 | |||
| GenBank mRNA | AK006942.1 | - | 4 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700072O05Rik| Gene Symbol: 1700072O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045733.1 | 4 | 630 | |||
| GenBank mRNA | AK006942.1 | - | 4 | 631 | ||