siRNA Details
| Gene Symbol: 1700072O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045733.1 | 4 | 656 | |||
| GenBank mRNA | AK006942.1 | - | 4 | 657 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706958.1 | - | 4 | 657 | Chr. 6 - 120554796 - 120574204 [+] on Build GRCm38 | |

1700072O05Rik
Genomic Map