siRNA Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050612.1 | - | 2 | 310 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB654278.1 | - | 2 | 306 | Chr. 17 - 32993132 - 32996970 [+] on Build GRCm38 | |

Zfp952
Genomic Map