siRNA Details
| Gene Symbol: Gm6416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046023.1 | 4 | 1366 | |||
| GenBank mRNA | AK041110.1 | - | 4 | 1370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6416| Gene Symbol: Gm6416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046023.1 | 4 | 1366 | |||
| GenBank mRNA | AK041110.1 | - | 4 | 1370 | ||