siRNA Details
| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110364.1 | 6 | 735 | |||
| NR_110365.1 | 6 | 672 | ||||
| GenBank mRNA | AK046045.1 | - | 6 | 733 | ||
| AK157435.1 | - | 6 | 671 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB312740.2 | - | 5 | 552 | Chr. 7 - 90036993 - 90049069 [+] on Build GRCm38 | |

E230029C05Rik
Genomic Map