siRNA Details
| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110364.1 | 6 | 642 | |||
| NR_110365.1 | 6 | 579 | ||||
| GenBank mRNA | AK046045.1 | - | 6 | 640 | ||
| AK157435.1 | - | 6 | 578 | |||
| BE650787.1 | - | 4 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230029C05Rik| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110364.1 | 6 | 642 | |||
| NR_110365.1 | 6 | 579 | ||||
| GenBank mRNA | AK046045.1 | - | 6 | 640 | ||
| AK157435.1 | - | 6 | 578 | |||
| BE650787.1 | - | 4 | 424 | |||