siRNA Details
| Gene Symbol: E230029C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110364.1 | 4, 5 | 456 | |||
| NR_110365.1 | 4, 5 | 361 | ||||
| GenBank mRNA | AK046045.1 | - | 4, 5 | 454 | ||
| AK157435.1 | - | 4, 5 | 360 | |||
| BE650787.1 | - | 2, 3 | 238 | |||

E230029C05Rik
Genomic Map