siRNA Details
| Gene Symbol: Gm16239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139491.1 | - | 1, 2 | 127 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725311.1 | - | 1, 2 | 123 | Chr. 10 - 99181645 - 99184607 [-] on Build GRCm38 | |

Gm16239
Genomic Map