siRNA Details
| Gene Symbol: 4930535L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019721.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715554.1 | - | 1 | 70 | Chr. 9 - 107998116 - 108002404 [-] on Build GRCm38 | |

4930535L15Rik
Genomic Map