siRNA Details
| Gene Symbol: 2900042G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019333.1 | - | 1 | 141 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712609.1 | - | 1 | 137 | Chr. 13 - 6530599 - 6531243 [+] on Build GRCm38 | |

2900042G08Rik
Genomic Map