siRNA Details
| Gene Symbol: H3f3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK161137.1 | - | 1 | 1216 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC027275.1 | - | 923 | Chr. - - [-] on Build | ||

H3f3a
Genomic Map