siRNA Details
| Gene Symbol: Rnf213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173199.1 | - | 3, 4 | 2303 | ||
| AK217856.1 | - | 3, 4 | 137 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI853801.1 | - | 3, 4 | 269 | Chr. 11 - 119468928 - 119474332 [+] on Build GRCm38 | |

Rnf213
Genomic Map