siRNA Details
| Gene Symbol: Gm2137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082723.1 | - | 1 | 1198 | ||
| BB396393.2 | - | 1 | 343 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM942997.1 | - | 1 | 192 | Chr. 10 - 61784310 - 61784805 [+] on Build GRCm38 | |

Gm2137
Genomic Map