siRNA Details
Gene Symbol: Gm2137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082723.1 | - | 1 | 1280 | ||
BB396393.2 | - | 1 | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM942997.1 | - | 1 | 274 | Chr. 10 - 61784310 - 61784805 [+] on Build GRCm38 |
