siRNA Details
| Gene Symbol: Immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006977.1 | - | 4 | 860 | ||
| AK164694.1 | - | 2 | 739 | |||
| BC145704.1 | - | 3 | 796 | |||
| BC145706.1 | - | 3 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Immt| Gene Symbol: Immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006977.1 | - | 4 | 860 | ||
| AK164694.1 | - | 2 | 739 | |||
| BC145704.1 | - | 3 | 796 | |||
| BC145706.1 | - | 3 | 796 | |||