siRNA Details
| Gene Symbol: 2810439L12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013270.1 | - | 1 | 263 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB797663.1 | - | 1 | 141 | Chr. 4 - 99655717 - 99655896 [-] on Build GRCm38 | |

2810439L12Rik
Genomic Map