siRNA Details
| Gene Symbol: 2210412B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008905.1 | - | 1 | 283 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708801.1 | - | 1 | 283 | Chr. 5 - 130131327 - 130131824 [+] on Build GRCm38 | |

2210412B16Rik
Genomic Map