siRNA Details
| Gene Symbol: 4930518C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015821.1 | - | 1 | 495 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715365.1 | - | 1 | 495 | Chr. 14 - 103899484 - 103900177 [+] on Build GRCm38 | |

4930518C04Rik
Genomic Map