siRNA Details
| Gene Symbol: 4930518C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015822.1 | - | 2 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715366.1 | - | 2 | 251 | Chr. 5 - 48003066 - 48005704 [-] on Build GRCm38 | |

4930518C09Rik
Genomic Map