siRNA Details
| Gene Symbol: 1010001B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003103.1 | - | 2 | 565 | ||
| AV005531.1 | - | 2 | 319 | |||
| Gene Symbol: Gm6566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042778.1 | - | 3 | 1148 | ||

1010001B22Rik
Genomic Map