siRNA Details
| Gene Symbol: 4930518I17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015828.1 | - | 1 | 466 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715371.1 | - | 1 | 466 | Chr. 15 - 32226385 - 32227040 [+] on Build GRCm38 | |

4930518I17Rik
Genomic Map