siRNA Details
| Gene Symbol: B230323A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040765.1 | 3 | 833 | |||
| GenBank mRNA | AK038443.1 | - | 3 | 834 | ||
| AK045913.1 | - | 1186 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230323A14Rik| Gene Symbol: B230323A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040765.1 | 3 | 833 | |||
| GenBank mRNA | AK038443.1 | - | 3 | 834 | ||
| AK045913.1 | - | 1186 | ||||