siRNA Details
| Gene Symbol: Gm19303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045092.1 | 6 | 1670 | |||
| GenBank mRNA | AK046060.1 | - | 6 | 1670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19303| Gene Symbol: Gm19303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045092.1 | 6 | 1670 | |||
| GenBank mRNA | AK046060.1 | - | 6 | 1670 | ||