siRNA Details
| Gene Symbol: 4933429O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033783.1 | 2 | 309 | |||
| GenBank mRNA | AK006575.1 | - | 2 | 311 | ||
| AK015198.1 | - | 2 | 265 | |||
| AK016985.1 | - | 2 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933429O19Rik| Gene Symbol: 4933429O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033783.1 | 2 | 309 | |||
| GenBank mRNA | AK006575.1 | - | 2 | 311 | ||
| AK015198.1 | - | 2 | 265 | |||
| AK016985.1 | - | 2 | 299 | |||