siRNA Details
| Gene Symbol: 9530051G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040272.1 | 4 | 1427 | |||
| GenBank mRNA | AK032014.1 | - | 3 | 1128 | ||
| AK035460.1 | - | 4 | 1428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530051G07Rik| Gene Symbol: 9530051G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040272.1 | 4 | 1427 | |||
| GenBank mRNA | AK032014.1 | - | 3 | 1128 | ||
| AK035460.1 | - | 4 | 1428 | |||