siRNA Details
| Gene Symbol: 9530052C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035466.1 | - | 2 | 1758 | ||
| AK085299.1 | - | 2 | 2037 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530052C20Rik| Gene Symbol: 9530052C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035466.1 | - | 2 | 1758 | ||
| AK085299.1 | - | 2 | 2037 | |||