siRNA Details
| Gene Symbol: Dbhos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040524.1 | 7 | 3421 | |||
| GenBank mRNA | AK032445.1 | - | 7 | 3423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbhos| Gene Symbol: Dbhos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040524.1 | 7 | 3421 | |||
| GenBank mRNA | AK032445.1 | - | 7 | 3423 | ||