siRNA Details
| Gene Symbol: Gm3805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006687.1 | - | 514 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706678.1 | - | 1 | 514 | Chr. 10 - 45577484 - 45578118 [-] on Build GRCm38 | |

Gm3805
Genomic Map