siRNA Details
| Gene Symbol: Gm5493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141778.1 | - | 227 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB655489.1 | - | 1 | 225 | Chr. 17 - 22739945 - 22750178 [+] on Build GRCm38 | |

Gm5493
Genomic Map