siRNA Details
| Gene Symbol: Gm30127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131720.1 | - | 2 | 258 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706382.1 | - | 2 | 258 | Chr. 13 - 35959157 - 35965008 [-] on Build GRCm38 | |

Gm30127
Genomic Map