siRNA Details
| Gene Symbol: 5830437K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017976.1 | - | 1 | 541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830437K03Rik| Gene Symbol: 5830437K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017976.1 | - | 1 | 541 | ||