siRNA Details
| Gene Symbol: Gm26908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134715.1 | - | 1166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm26908| Gene Symbol: Gm26908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134715.1 | - | 1166 | |||