siRNA Details
| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102296.1 | 3, 4 | 377 | |||
| GenBank mRNA | AK049386.1 | - | 3, 4 | 365 | ||
| AK089981.1 | - | 367 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C430002N11Rik| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102296.1 | 3, 4 | 377 | |||
| GenBank mRNA | AK049386.1 | - | 3, 4 | 365 | ||
| AK089981.1 | - | 367 | ||||