siRNA Details
| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102296.1 | 4 | 403 | |||
| GenBank mRNA | AK049386.1 | - | 4 | 391 | ||
| AK089981.1 | - | 393 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB652180.1 | - | 4 | 391 | Chr. 9 - 96765575 - 96773390 [+] on Build GRCm38 | |

C430002N11Rik
Genomic Map