siRNA Details
| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102293.1 | 1, 2 | 115 | |||
| NR_102296.1 | 1, 2 | 115 | ||||
| GenBank mRNA | AA030139.1 | - | 1, 2 | 76 | ||
| AK049386.1 | - | 1, 2 | 103 | |||
| AK089981.1 | - | 105 | ||||

C430002N11Rik
Genomic Map