siRNA Details
| Gene Symbol: Gm33474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084468.1 | - | 2 | 305 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB658656.1 | - | 2 | 301 | Chr. 5 - 107048839 - 107055428 [+] on Build GRCm38 | |

Gm33474
Genomic Map