siRNA Details
Gene Symbol: C030037F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021128.1 | - | 1 | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728262.1 | - | 1 | 106 | Chr. 8 - 14745320 - 14747427 [-] on Build GRCm38 |
